Learn More
Chemglass Life Sciences 50/30 O-Ring Ball Joint, Clamp Size:50, O-Ring Size: 128, Tubing OD: 36mm
SDP
Catalog No. CG12210
Encompass_Preferred
50/30 O-Ring Ball Joint, Clamp Size:50, O-Ring Size: 128,Tubing OD: 36mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12210 Supplier Chemglass Life Sciences Supplier No. CG12210
50/30 O-Ring Ball Joint, Clamp Size:50, O-Ring Size: 128,Tubing OD: 36mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.