Learn More
Chemglass Life Sciences Joint, 50/30, Socket, Clamp Size 50, 36mm Tubing
SDP
Catalog No. CG12112
Encompass_Preferred
Joint, 50/30, Socket, Clamp Size 50, 36mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12112 Supplier Chemglass Life Sciences Supplier No. CG12112
Joint, 50/30, Socket, Clamp Size 50, 36mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.