Learn More
Chemglass Life Sciences 50/50 Outer Joint, Full Length, on 54mm OD x 2.4mm W tubing
SDP
Catalog No. CG10111
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 50/50 Outer Joint, Full Length, on 54mm OD x 2.4mm W tubing.
Catalog No. CG10111 Supplier Chemglass Life Sciences Supplier No. CG10111
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 50/50 Outer Joint, Full Length, on 54mm OD x 2.4mm W tubing.