Learn More
Exel International 50 to 60cc Syringes
Description
Latex free;Sterile;Non-toxic;Pyrogen free;Positive plunger stop to prevent spillage when drawing;Smooth plunger motion;Polypropylene construction.
Specifications
Specifications
| Certifications/Compliance | Pyrogen Free |
| Material | Polypropylene |
| Latex Free | Yes |
| Product Type | Syringe |
| Volume (Metric) | 50 to 60 mL |
| Connection Type | Catheter |
| Graduated | Yes |
| Sterility | Non-sterile |