Learn More
Thermco Products Inc -50/70C TRIPLE DIS AMBIENT
Catalog No. NC0910002
Encompass_Preferred
Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible alarm will sound when the temperature rises above or goes below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend. Reset buttons on front panel. Each thermometer is serialized and traceable to NIST.The thermometer probe is housed in an unbreakable plastic bottle or vial filled with a Bio-Safe Glycol / Water or Sand buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets allowing attachment to doors or walls of the instrument. Other features include a flip-open stand, wall mounting key hole, and supplied with a AAA alkaline battery. The instrument is �C and �F switchable.
Catalog No. NC0910002 Supplier Thermco Products Inc Supplier No. ACC895AMB
Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible alarm will sound when the temperature rises above or goes below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend. Reset buttons on front panel. Each thermometer is serialized and traceable to NIST.The thermometer probe is housed in an unbreakable plastic bottle or vial filled with a Bio-Safe Glycol / Water or Sand buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets allowing attachment to doors or walls of the instrument. Other features include a flip-open stand, wall mounting key hole, and supplied with a AAA alkaline battery. The instrument is �C and �F switchable.