Learn More
DuPont™ ProShield™ 50 Bib Apron NB273B WH
Catalog No. 17897237 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Universal
Catalog No. Size
17-897-237 Universal
1 options
Catalog No. 17-897-237 Supplier DuPont™ ProShield™ Supplier No. NB273BWH00010000
  • $252.31
    $199.65 / Case of 100
    Save $52.66
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
50 Bib Apron with neck loop, waist ties, bound seams, and durable design for dependable industrial and sanitation protection.
  • ProShield™ 50 garments provide a barrier against non-hazardous aerosols, light liquid splash, and dry particles
  • Lightweight design allows comfort and mobility during extended use
  • Enhanced frontal protection for industrial and sanitation tasks
  • Bound seams around the neck for added durability
  • Ties at the waist ensure secure fit
  • Manufactured without natural rubber latex
  • One size fits most for versatile use
TRUSTED_SUSTAINABILITY
Specifications
Type Bib Apron
Size Universal
Material Polyethylene Laminate
Color White
Disposable Single-use
Closure Type Neck Loop, Waist Ties
Sterility Non-sterile
No. of Pockets None
Dimensions (L x W) 35.75 x 27 in.
Seam Type Bound