Learn More
Alkali Scientific CellPro™ Bottle Top Filtration Filters, PES Membrane, 50mL, 0.22µm, Individually Wrapped, Sterile, 24/cs
SDP
Supplier:  Alkali Scientific VH05022
Encompass_Preferred
CellPro™ Bottle Top Filters are available in 0.22µm PES membrane. The screw-on funnel includes a lid and a graduated nozzle connection to fit multiple diameter vacuum hoses. They also feature engraved graduation marks to ensure accuracy and are gamma sterilized and certified RNAse, DNAse, and pyrogen free.
Catalog No. 50-233-5971