Learn More
Gold Biotechnology Inc 50 bp DNA Ladder 500 µL
SDP
Catalog No. 501532762
Encompass_Preferred
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 17 fragments, suitable for gel electrophoresis. The DNA includes fragments ranging from 50-1,500 base pairs. The 200 and 500 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 g a load) for approximating the mass of DNA in comparably intense samples of a similar size. The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.
Catalog No. 50-153-2762 Supplier Gold Biotechnology Inc Supplier No. D100500
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 17 fragments, suitable for gel electrophoresis. The DNA includes fragments ranging from 50-1,500 base pairs. The 200 and 500 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 g a load) for approximating the mass of DNA in comparably intense samples of a similar size. The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.