Learn More
Medchemexpress LLC 50 bp DNA Marker | 1 ML
SDP
Supplier:  Medchemexpress LLC HYK08011ML
Encompass_Preferred
The 50 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer, which can be directly used for nucleic acid electrophoresis analysis. The Marker contains 14 double-stranded DNA fragments ranging from 50 bp to 1000 bp. 5 μL of this product contains about 100 ng for the 250 or 500 bp bands, and about 40 ng for the other bands.
  • Can be directly used for nucleic acid electrophoresis analysis.
  • Contains 14 double-stranded DNA fragments ranging from 50 bp to 1000 bp.
Catalog No. 50-004-31533