Learn More
Wesco Industrial Products 50BTW Series Drum Truck
Catalog No. NC0777302 Shop All Wesco Industrial Products Products
Encompass_Preferred
Poly Drum Truck & DIspensing 50BTW - Hard Polyolefin Wheels Acts as a Poly Drum Truck or Drain Stand. Wide “feet” and belt eliminate the risk of drums being punctured by nose prongs and chime hooks. With two wheels resting on floor, 32" to center of belt. Handles all poly drums except overpacks). Handles 30, 55 & 85 gal. drums. Steel, poly & fiber drums. Capacity 1000 lbs. Weight 65 lbs.
Catalog No. NC0777302 Supplier Wesco Industrial Products Supplier No. 240078
Poly Drum Truck & DIspensing 50BTW - Hard Polyolefin Wheels Acts as a Poly Drum Truck or Drain Stand. Wide “feet” and belt eliminate the risk of drums being punctured by nose prongs and chime hooks. With two wheels resting on floor, 32" to center of belt. Handles all poly drums except overpacks). Handles 30, 55 & 85 gal. drums. Steel, poly & fiber drums. Capacity 1000 lbs. Weight 65 lbs.