Learn More
Alkali Scientific ASI™ 50 Bulk Beads, Stainless Steel, 5mm, alcohol washed, 1000/pk
SDP
Catalog No. 503028345
Encompass_Preferred
5mm diameter steel beads are suitable for homogenization of tough, hard samples, plant materials, seeds and dried samples. Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free. Can be used with any bead homogenizer. 1000 beads per pack.
Catalog No. 50-302-8345 Supplier Alkali Scientific Supplier No. RS113450
5mm diameter steel beads are suitable for homogenization of tough, hard samples, plant materials, seeds and dried samples. Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free. Can be used with any bead homogenizer. 1000 beads per pack.