Learn More
Alkali Scientific 50mL Centrifuge Tubes, Conical, Bagged, Sterile, Blue Cap, PP, 500/cs
SDP
Catalog No. 502336105
Encompass_Preferred
50mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free
Catalog No. 50-233-6105 Supplier Alkali Scientific Supplier No. C5605
50mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free