Learn More
Helix Medical Inc 50'COIL ID: 0.500 OD: 0.750 .2
Catalog No. NC1125901
Encompass_Preferred
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations.
Catalog No. NC1125901 Supplier Helix Medical Inc Supplier No. 6001133
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations.