Learn More
Saint Gobain Performance Plastics 50 FT TUBING 3/8 5/8 1/8 50/PK
Supplier:  Saint Gobain Performance Plastics ACF00029F
Encompass_Preferred
Saint-GobainTygon S3 E-3603 Flexible Tubings Diameter Inner 3/8 in. Diameter Outer 5/8 in. Thickness 1/8 in. Overall Length 50 ft. 15 m
Catalog No. 50-206-8957