Learn More
Teknova 50% (v/v) Glycerol Solution. 500mL, Sterile.
SDP
Supplier:  Teknova G1798
Encompass
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.
Catalog No. 50-841-704