Learn More
Analytical Sales & Services 50µL PEEK Sample Loop
Catalog No. 5000805538
Encompass
50µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-05538 Supplier Analytical Sales & Services Supplier No. 490550234
50µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH