Learn More
Ace Glass, Inc. Spherical flask, 50000mL, (3)in-line 65/40 spherical joints, (2)24/40 standard taper joints
SDP
Catalog No. 50978752
Encompass
50L spherical flask with (3) in-line 65/40 spherical joints, and (2) 24/40 standard taper joints. Flask is designed for use with ACE 50L Spherical Reaction Assembly.
Catalog No. 50-978-752 Supplier Ace Glass, Inc. Supplier No. 647026
50L spherical flask with (3) in-line 65/40 spherical joints, and (2) 24/40 standard taper joints. Flask is designed for use with ACE 50L Spherical Reaction Assembly.