Learn More
Polyscience 50 Micron Sediment Filter Kit - Without Bypass
SDP
Supplier:  Polyscience 510519
Encompass_Preferred
PolyScience™ 50 Micron Sediment Filter Kit without BypassFor use with PolyScience LS Series, LM Series and MM Series Benchtop Chillers Package of 1
Catalog No. NC1493496