Learn More
Labcon North America 50mL PerformR Freestanding Centrifuge Tubes, in Bulk
Catalog No. NC0572006
Encompass_Preferred
Tube, Freestanding conical centrifuge; Labcon; PerformR; 50mL; With screw caps in separate bag, black side graduations, and white writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable to -80C; Tubes are autoclavable; Capacity: 50mL; Teal color caps; 500 tubes and 500 caps in separate bags; Made in USA
Catalog No. NC0572006 Supplier Labcon North America Supplier No. 30958700089
Tube, Freestanding conical centrifuge; Labcon; PerformR; 50mL; With screw caps in separate bag, black side graduations, and white writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable to -80C; Tubes are autoclavable; Capacity: 50mL; Teal color caps; 500 tubes and 500 caps in separate bags; Made in USA