Learn More
Anton Paar Pack of 500 polypropylene vials 50ml
Supplier:  Anton Paar 227935
Encompass_Preferred
One package including 500 pieces of 50 mL PP vials for open sample digestion and extraction.Suitable for use with Anton Paar Multicube 48 Hot block digestion heating unit NC1818247
Catalog No. NC1818258