Learn More
Anton Paar Pack of 500 polypropylene watch glasses for 50ml vials
Supplier:  Anton Paar 227934
Encompass_Preferred
One package including 500 pieces of watch glasses for covering 50 mL PP vials during digestion and extraction.Suitable for use with Anton Paar Multicube 48 Hot block digestion heating unit NC1818247
Catalog No. NC1818263