Learn More
SAI Infusion Technologies 3D Mini & MiniBT Reservoirs - 50mL
SDP
Catalog No. NC0213999
Encompass
50mL syringe reservoirs for use with the SAI 3D Mini Ambulatory Pump or the SAI 3D MiniBT Ambulatory pump. 3D Mini pumps are lightweight and excellent for ambulatory infusion studies in primates and other large animals. Reservoirs are used to refill infusion solutions during infusion studies. 60/pk. Product is for laboratory animal research only.
Catalog No. NC0213999 Supplier SAI Infusion Technologies Supplier No. CRN50
50mL syringe reservoirs for use with the SAI 3D Mini Ambulatory Pump or the SAI 3D MiniBT Ambulatory pump. 3D Mini pumps are lightweight and excellent for ambulatory infusion studies in primates and other large animals. Reservoirs are used to refill infusion solutions during infusion studies. 60/pk. Product is for laboratory animal research only.