Learn More
Waters Corp Tube Cooled Domino, 50 mL
Supplier: Waters Corp 186010158
This machine-readable consumable holder is designed to interface with the Andrew+ pipetting robot. It can hold specific 50 mL tubes and its cooled design aids in sample preservation.
- Machine-readable consumable holder
- Designed to interface with the Andrew+ pipetting robot
- Holds specific 50 mL tubes
- Cooled design for sample preservation