Learn More
Perkin Elmer US LLC 50 mm, 3.0 mm I.D., 1.7 Quasar AQ LC Column
Supplier:  Perkin Elmer US LLC N9308856
Encompass_Preferred
Quasar AQ LC columns benefit from no phase collapse even in 100% aqueous mobile phase conditions and the polar end capping aids in retention of more polar compounds.
Catalog No. 50-211-3029