Learn More
ATHENA ES 50 mM a-cyclodextrin
Supplier:  ATHENA ES 0611
Encompass_Preferred
Cyclodextrins’ unique molecular structure greatly promotes protein refolding. Cyclodextrins can suppress protein aggregation, protect against degradation and loss of protein function. They also bind well to detergents (aggregation suppressors), which allows for easy separation of the detergent from the protein, encouraging proper refolding. Differentcyclodextrins act and respond differently to different protein structures. Because of this, AthenaESTM provides 3 of the most common cyclodextrins in a simple screening kit so that the ideal conditions for refolding a given protein can be found quickly and easily. The Cyclodextrin Screening Kit can be particularly useful when paired with the Detergent Screening Kit. Individual cyclodextrins are also available for purchase.
Catalog No. 50-001-58075