Learn More
Nanocomposix NanoXact Silica Nanospheres - Aminated - 50 nm, 10 mg/mL in ethanol, 25 mL
SDP
Catalog No. NC1251771
Encompass_Preferred
As a global leader in silica nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.50 nm spherical silica nanoparticles combine biocompatibility, unique optical properties, and versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from nanomedicine, targeted drug delivery, UV-scattering, catalysis, DNA/RNA extraction, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC1251771 Supplier Nanocomposix Supplier No. SIAN5025M
As a global leader in silica nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.50 nm spherical silica nanoparticles combine biocompatibility, unique optical properties, and versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from nanomedicine, targeted drug delivery, UV-scattering, catalysis, DNA/RNA extraction, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.