Learn More
Andwin Scientific 50-PLACE CRYOVIAL WORKSTATION
SDP
Catalog No. 50819936
Encompass_Preferred
The 50-place Cryovial rack measures 4" W x 8" L x 1" H. It is constructed with polypropylene. This is sold in a pack of 2.
Catalog No. 50-819-936 Supplier Andwin Scientific Supplier No. 23315200
The 50-place Cryovial rack measures 4" W x 8" L x 1" H. It is constructed with polypropylene. This is sold in a pack of 2.