Learn More
Electron Microscopy Sciences 50 Place Scintillation Vial Rack, White
SDP
Catalog No. 50365978
Encompass_Preferred
Rack measures: 12 3/4" x 6 3/4" x 1 3/8" high and has 50 cavities, 1.1" diameter x .75" deep, cavities have a center drain hole 1/2" in diameter. The racks are stackable when not in use and are steam autoclavable at 250ºF.
Catalog No. 50-365-978 Supplier Electron Microscopy Sciences Supplier No. 72635
Rack measures: 12 3/4" x 6 3/4" x 1 3/8" high and has 50 cavities, 1.1" diameter x .75" deep, cavities have a center drain hole 1/2" in diameter. The racks are stackable when not in use and are steam autoclavable at 250ºF.