Learn More
DuPont™ ProShield™ 50 Sleeve NB500S WH
Catalog No. 17897238 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Universal
Catalog No. Size
17-897-238 Universal
1 options
Catalog No. 17-897-238 Supplier DuPont™ ProShield™ Supplier No. NB500SWH000200YU
  • $217.50
    $149.65 / Case of 200
    Save $67.85
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
50 Sleeve with elastic ends and serged seams for durable, reliable arm protection in industrial and sanitation use.
  • ProShield™ 50 microporous film garments protect against non-hazardous aerosols, light liquid splashes, and dry particles
  • Lightweight and roomy design for comfort and ease of movement
  • Ideal for janitorial, sanitation, industrial maintenance, asbestos abatement, and hazardous remediation
  • Elastic at both ends for secure arm protection
  • Manufactured without natural rubber latex
  • One size fits most
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Non-woven Spunbond Polypropylene, Polyethylene Laminate Film
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Latex Free No
Length (English) 24 in.
Length (Metric) 47.62 cm