Learn More
Electron Microscopy Sciences Unit Staining Dish
SDP
Catalog No. 5028700
Encompass_Preferred
Slide staining set contains a nontarnishable stainless steel, slotted rack, designed to hold 50 slides in a variety of sizes( 3" x 1", 3" x 1.5", and 3" x 2"), a dish and cover. The unit measures approximately 200 x 105 x 83 mm overall. The rack is resistant to most staining solutions. The handle is permanently attached allowing for closure of the dish and for easy insertion and removal of the slides.
Catalog No. 50-287-00 Supplier Electron Microscopy Sciences Supplier No. 7031251
Slide staining set contains a nontarnishable stainless steel, slotted rack, designed to hold 50 slides in a variety of sizes( 3" x 1", 3" x 1.5", and 3" x 2"), a dish and cover. The unit measures approximately 200 x 105 x 83 mm overall. The rack is resistant to most staining solutions. The handle is permanently attached allowing for closure of the dish and for easy insertion and removal of the slides.