Learn More
Electron Microscopy Sciences 50 Small bags for Lab-sorb
SDP
Catalog No. 5034512
Encompass_Preferred
Extra durable bags (500-600ml) for use with the Lab-Sorb granules to clean up spills.
Catalog No. 50-345-12 Supplier Electron Microscopy Sciences Supplier No. 6855211
Extra durable bags (500-600ml) for use with the Lab-Sorb granules to clean up spills.