Learn More
ACROBiosystems Recombinant Protein;Human FcRn / FCGRT & B2M Heterodimer Protein (SPR verified);HEK293;50UG;FCM,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems FCMH528650UG
Encompass_Preferred
Recombinant Protein;Human FcRn / FCGRT & B2M Heterodimer Protein (SPR verified);50UG;Human FcRn Heterodimer Protein (SPR verified) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # NP_001129491.1 (FCGRT) & AAH32589 (B2M)).;Human FcRn Heterodimer Protein (SPR verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 31.2 kDa (FCGRT) and 13.1 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) is fused with Strep II-tag at the C-terminus. The reducing (R) protein migrates as 33 kDa (FCGRT) and 13 kDa (B2M) respectively due to glycosylation.;Immobilized Ipilimumab at 10 μg/mL (100 μL/well) can bind Human FcRn Heterodimer Protein (SPR verified) (Cat. No. FCM-H5286) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Human FcRn Heterodimer Protein (Cat. No. FCM-H5286) on CM5 Chip via anti-His antibody, can bind Herceptin wit
Catalog No. NC0843097