Learn More
Chemglass Life Sciences Chemglass Nest 50 μL Automation Conductive Filter Tips for Hamilton Systems, Sterile, 5 x 96-Count Blister Packs (5760/cs)
SDP
Catalog No. 502733278
Encompass_Preferred
Chemglass Nest 50 μL automation conductive filter tips are designed for automated liquid-handling on Hamilton systems. Sterile, barcode-labeled blister packs provide traceability and reduce contamination risk in high-throughput workflows such as genomics, proteomics, and biopharmaceutical assays.
  • 50 μL nominal volume
  • Sterile; electron beam sterilized
  • Medical-grade polypropylene construction
  • DNase- and RNase-free
  • Aerosol-resistant filter to reduce contamination
  • Barcode-labeled blister packs for traceability
  • Packaged as five blister packs of 96 tips each (5760 tips per case)
Catalog No. 50-273-3278 Supplier Chemglass Life Sciences Supplier No. CLS3499A021
Chemglass Nest 50 μL automation conductive filter tips are designed for automated liquid-handling on Hamilton systems. Sterile, barcode-labeled blister packs provide traceability and reduce contamination risk in high-throughput workflows such as genomics, proteomics, and biopharmaceutical assays.
  • 50 μL nominal volume
  • Sterile; electron beam sterilized
  • Medical-grade polypropylene construction
  • DNase- and RNase-free
  • Aerosol-resistant filter to reduce contamination
  • Barcode-labeled blister packs for traceability
  • Packaged as five blister packs of 96 tips each (5760 tips per case)