Learn More
Chemglass Life Sciences Chemglass 50 μL Automation Conductive Pipette Tips for Hamilton systems, sterile, barcode, 96 x 5 blister pack
SDP
Catalog No. 502733277
Encompass_Preferred
Sterile, conductive 50 μL automation pipette tips compatible with Hamilton automated liquid handling systems. Preloaded in barcode-labelled blister packs for automated loading and inventory tracking, these tips are designed for high-throughput applications and are RNase/DNase-free.
  • 50 μL volume
  • Medical-grade polypropylene
  • Sterile; electron-beam sterilized
  • RNase/DNase free
  • Conductive for use with liquid level sensing
  • Barcode on blister packs for automation and inventory
  • Packaged as 96 tips per blister, 5 blisters per pack (5,760 tips per case)
  • Preloaded in blister packs for automated loading
  • Produced in a controlled clean room environment
Catalog No. 50-273-3277 Supplier Chemglass Life Sciences Supplier No. CLS3499A022
Add to Cart
Add to Cart
Sterile, conductive 50 μL automation pipette tips compatible with Hamilton automated liquid handling systems. Preloaded in barcode-labelled blister packs for automated loading and inventory tracking, these tips are designed for high-throughput applications and are RNase/DNase-free.
  • 50 μL volume
  • Medical-grade polypropylene
  • Sterile; electron-beam sterilized
  • RNase/DNase free
  • Conductive for use with liquid level sensing
  • Barcode on blister packs for automation and inventory
  • Packaged as 96 tips per blister, 5 blisters per pack (5,760 tips per case)
  • Preloaded in blister packs for automated loading
  • Produced in a controlled clean room environment