Learn More
USA Scientific Inc 50 uL black tip for Tecan
Catalog No. 502745541
Encompass
TipOne for Tecan Genesis and Freedom EVO. 50 uL conductive black tips, 12 twin trays of 96 tips (2304 tips)
Catalog No. 50-274-5541 Supplier USA Scientific Inc Supplier No. 11815709
TipOne for Tecan Genesis and Freedom EVO. 50 uL conductive black tips, 12 twin trays of 96 tips (2304 tips)