Learn More
Starna Cells Inc 50 UM CUVETTE
SDP
Catalog No. NC9234251
Encompass_Preferred
Fluorimeter, Sub-micro, Square open top, Spectrosil quartz, 2W x 2.5H, 12.5L x 12.5W x 45H, 0.050mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal
Catalog No. NC9234251 Supplier Starna Cells Inc Supplier No. 16.50FQ10/Z15
Fluorimeter, Sub-micro, Square open top, Spectrosil quartz, 2W x 2.5H, 12.5L x 12.5W x 45H, 0.050mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal