Learn More
Molecular Dimensions 50% V/V PEG SMEAR LOW 250ML
SDP
Catalog No. NC2064009
Encompass
PEG is an effective crystallization agent due to its protein-protein interaction promoting properties. The BCS Screen overcomes the impracticality of sampling all PEG varieties that has led to a few variants predominating in the most common screens. PEG Smears are designed primarily for efficient screening of chemical space. It is generally thought that well-defined, single PEG species produce the best diffracting crystals. Thus, optimization might involve identifying the single PEG compound that produces the best crystal for structural studies. MOLECULAR DIMENSIONS INC mfg# MD2-250-258
Catalog No. NC2064009 Supplier Molecular Dimensions Supplier No. MD2250258
PEG is an effective crystallization agent due to its protein-protein interaction promoting properties. The BCS Screen overcomes the impracticality of sampling all PEG varieties that has led to a few variants predominating in the most common screens. PEG Smears are designed primarily for efficient screening of chemical space. It is generally thought that well-defined, single PEG species produce the best diffracting crystals. Thus, optimization might involve identifying the single PEG compound that produces the best crystal for structural studies. MOLECULAR DIMENSIONS INC mfg# MD2-250-258