Learn More
Sigma Aldrich Fine Chemicals Biosciences 50-Well microtube storage
Catalog No. 502900903
Encompass
Ideal for sample or case study storage.Made from polypropyleneGreat for smaller storage requirements of 1.5/2.0 mL microtubes or cryogenic vialsEasy-open hinged lid with snap claspTubes 1.5/2.0 mLWells 50Well Dia 12.3 mmDimensions 5.6 x 3.6 x 2.2 in (14.2 x 9.2 x 5.6 cm)Packaging 5/pk Autoclavable
Catalog No. 50-290-0903 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120033
Ideal for sample or case study storage.Made from polypropyleneGreat for smaller storage requirements of 1.5/2.0 mL microtubes or cryogenic vialsEasy-open hinged lid with snap claspTubes 1.5/2.0 mLWells 50Well Dia 12.3 mmDimensions 5.6 x 3.6 x 2.2 in (14.2 x 9.2 x 5.6 cm)Packaging 5/pk Autoclavable