Learn More
Saint Gobain Performance Plastics .50 x .75 x 15 BRAIDED TUBING1PK
Catalog No. 502069047 Shop All Saint Gobain Performance Plastics Products
Encompass_Preferred
Saint-Gobain C-Flex Tubing 082 Formulation - Braided Diameter Inner 1/2 in. 12.7 mm Diameter Outer 3/4 in. 19.1 mm Overall Length 50 ft. 15.2 m Autoclavable Autoclavable Certifications/Compliance USP Class VI ISO 10993 EP Flexibility Flexible Hardness 60 (Shore A)
Catalog No. 50-206-9047 Supplier Saint Gobain Performance Plastics Supplier No. CFB0825004
Saint-Gobain C-Flex Tubing 082 Formulation - Braided Diameter Inner 1/2 in. 12.7 mm Diameter Outer 3/4 in. 19.1 mm Overall Length 50 ft. 15.2 m Autoclavable Autoclavable Certifications/Compliance USP Class VI ISO 10993 EP Flexibility Flexible Hardness 60 (Shore A)