Learn More
Accuflow Systems Saint-Gobain C-Flex TPE Tubing, 1/2 in x 3/4 in, Translucent
Supplier:  Accuflow Systems 0825004M
Encompass_Preferred
C-Flex 082 TPE braided tubing designed for biopharmaceutical fluid transfer, offering weldable and sealable connections and validated bioprocess compatibility.
  • Inner diameter 0.500 in (12.7 mm)
  • Outer diameter 0.750 in (19.1 mm)
  • Material: thermoplastic elastomer (C-Flex 082)
  • Translucent, non-pyrogenic, non-cytotoxic
  • Sealable and weldable pre- or post-sterilization; compatible with autoclave and gamma irradiation
  • Biopharmaceutical grade with validation packages available; USP Class VI compliance
Catalog No. NC2312786