Learn More
Accuflow Systems Saint-Gobain C-Flex TPE Tubing, 0.5 in ID, 0.75 in OD, 50 ft Spool
Catalog No. NC2312795
Encompass_Preferred
Saint-Gobain C-Flex TPE Tubing, Formulation 374, is a translucent thermoplastic elastomer tubing designed for high-purity biopharmaceutical and laboratory fluid transfer. This formulation provides heat-seal and sterile-weld capability, is compatible with peristaltic pumps, and is suitable for single-use assemblies and aseptic fluid handling.
  • Translucent thermoplastic elastomer for visual flow monitoring.
  • 0.500 in ID x 0.750 in OD, 0.125 in wall thickness.
  • 50 ft (15 m) spool length.
  • Sterilizable by gamma and X-ray irradiation up to 50 kGy; one-time autoclave at 121 °C for 30 minutes.
  • Compatible with peristaltic pumps and single-use fluid transfer assemblies.
  • Provides heat-seal and sterile-weld performance for aseptic connections.
  • Suitable for buffer and media handling, cell culture, and aseptic welding operations.
Catalog No. NC2312795 Supplier Accuflow Systems Supplier No. 3745004M
Saint-Gobain C-Flex TPE Tubing, Formulation 374, is a translucent thermoplastic elastomer tubing designed for high-purity biopharmaceutical and laboratory fluid transfer. This formulation provides heat-seal and sterile-weld capability, is compatible with peristaltic pumps, and is suitable for single-use assemblies and aseptic fluid handling.
  • Translucent thermoplastic elastomer for visual flow monitoring.
  • 0.500 in ID x 0.750 in OD, 0.125 in wall thickness.
  • 50 ft (15 m) spool length.
  • Sterilizable by gamma and X-ray irradiation up to 50 kGy; one-time autoclave at 121 °C for 30 minutes.
  • Compatible with peristaltic pumps and single-use fluid transfer assemblies.
  • Provides heat-seal and sterile-weld performance for aseptic connections.
  • Suitable for buffer and media handling, cell culture, and aseptic welding operations.