Learn More
Medchemexpress LLC 500 BP DNA MARKER 1ML | 1 ML
SDP
Catalog No. 5000430190
Encompass_Preferred
The 500 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer and can be directly used for nucleic acid electrophoresis analysis. It contains 14 double-stranded DNA fragments ranging from 500 bp to 5000 bp.
  • Contains 14 double-stranded DNA fragments.
  • Suitable for nucleic acid electrophoresis analysis.
  • Each 5 μL contains approximately 100 ng for the 2500 bp band and 40 ng for other bands.
Catalog No. 50-004-30190 Supplier Medchemexpress LLC Supplier No. HYK08041ML
The 500 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer and can be directly used for nucleic acid electrophoresis analysis. It contains 14 double-stranded DNA fragments ranging from 500 bp to 5000 bp.
  • Contains 14 double-stranded DNA fragments.
  • Suitable for nucleic acid electrophoresis analysis.
  • Each 5 μL contains approximately 100 ng for the 2500 bp band and 40 ng for other bands.