Learn More
Analytical Sales & Services 500µL 96-Well Collection Plate with Wide Mouth, Round Bottom Wells
Catalog No. 5000804211
Encompass
Advantage™ 500µL 96-Well Collection Plate\n• Wide Mouth Round Wells with Round Bottoms\n• Polypropylene\n\nQty/Pkg: 50\n\nCompatible with the wide mouth cap mats (955011, 955022, 966011, 966022)\n
\n
Catalog No. 50-008-04211 Supplier Analytical Sales & Services Supplier No. 96500
Advantage™ 500µL 96-Well Collection Plate\n• Wide Mouth Round Wells with Round Bottoms\n• Polypropylene\n\nQty/Pkg: 50\n\nCompatible with the wide mouth cap mats (955011, 955022, 966011, 966022)\n
\n