Learn More
Analytical Sales & Services 500µL GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000804303
Encompass
500µL GC Syringe 1750N for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-04303 Supplier Analytical Sales & Services Supplier No. 42030804
500µL GC Syringe 1750N for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH