Learn More
Analytical Sales & Services 500µL PEEK Sample Loop
Catalog No. 5000805689
Encompass
500µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-05689 Supplier Analytical Sales & Services Supplier No. 490550264
Add to Cart
Add to Cart
500µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH