Learn More
Analytical Sales & Services 500µL Removable Needle Syringe
Catalog No. 5000804912
Encompass
500µL Removable Needle Syringe, 22s Gauge, Point Style 3
Qty/Pkg: EACH
Catalog No. 50-008-04912 Supplier Analytical Sales & Services Supplier No. 4808654
500µL Removable Needle Syringe, 22s Gauge, Point Style 3
Qty/Pkg: EACH