Learn More
Perkin Elmer US LLC 500 µL Syringe Kit with 1000 µL Buffer Tubing
Catalog No. 502112571
Encompass_Preferred
These glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.
Catalog No. 50-211-2571 Supplier Perkin Elmer US LLC Supplier No. N2936053
These glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.