Learn More
Andwin Scientific 500 ML BUFFER PH 7.00
SDP
Catalog No. NC9501891
Encompass_Preferred
The Standard Buffer Solution is a compound made to the specific pH value of +6.99 to 7.01 This product helps in resisting the change in pH when moderate amounts of either a base or acid are added, or on dilution with a solvent. This is sold in the quantity of 500 mL.
Catalog No. NC9501891 Supplier Andwin Scientific Supplier No. 34170115
The Standard Buffer Solution is a compound made to the specific pH value of +6.99 to 7.01 This product helps in resisting the change in pH when moderate amounts of either a base or acid are added, or on dilution with a solvent. This is sold in the quantity of 500 mL.