Learn More
Alkali Scientific 500 mL Erlenmeyer Flask Bi-dir
SDP
Catalog No. 503028692
Encompass_Preferred
Bi-directional Transfer Cap for 125-1000mL Erlenmeyer Flask is equipped with the Filter of 0 2 m PVDF Membrane And the pipe for liquid transfer is outfitted with Male MPC Connector or the Luer Taper and the corresponding Seal Cap which makes it convenient for customers to connect the ends of pipes with the same connector Besides the tube at the bottom can reach the flask bottom then completely and aseptically transfer the liquid Meanwhile contamination risk is greatly reduced due to the simplified aseptic transfer procedures with the Bi-directional Transfer Cap for Erlenmeyer Flask Product Features Closed Liquid Transfer reduces the contamination risk during the operation procedures The vent cap with 0 2um membrane is permeable and waterproof thus can effectively block the microorganisms and avoids the contamination but ensures the gas exchange One end of the pipe is outfitted with male MPC Connector or Luer Taper making the operation safer and more convenient The close connect
Catalog No. 50-302-8692 Supplier Alkali Scientific Supplier No. EFC404
Bi-directional Transfer Cap for 125-1000mL Erlenmeyer Flask is equipped with the Filter of 0 2 m PVDF Membrane And the pipe for liquid transfer is outfitted with Male MPC Connector or the Luer Taper and the corresponding Seal Cap which makes it convenient for customers to connect the ends of pipes with the same connector Besides the tube at the bottom can reach the flask bottom then completely and aseptically transfer the liquid Meanwhile contamination risk is greatly reduced due to the simplified aseptic transfer procedures with the Bi-directional Transfer Cap for Erlenmeyer Flask Product Features Closed Liquid Transfer reduces the contamination risk during the operation procedures The vent cap with 0 2um membrane is permeable and waterproof thus can effectively block the microorganisms and avoids the contamination but ensures the gas exchange One end of the pipe is outfitted with male MPC Connector or Luer Taper making the operation safer and more convenient The close connect