Learn More
Bellco Glass, Inc. 500 mL PC Conical Erlenmeyer Flasks, with Baffles, Seal Caps, Sterile,1/pk, 12/cs
SDP
Catalog No. 502329718
Encompass_Preferred
500 mL PC Conical Erlenmeyer Flasks, with Baffles, Seal Caps, Sterile,1/pk, 12/cs. Ideal for shaker culture applications. There are two kinds of bottle materials, both PETG and PC are available. PC bottle or PEFG bottleSterilized by E-BeamNonpyrogenicIndividually wrapped in a plastic bag.Both vent caps and plug seal caps are available.Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.
Catalog No. 50-232-9718 Supplier Bellco Glass, Inc. Supplier No. 451400196
500 mL PC Conical Erlenmeyer Flasks, with Baffles, Seal Caps, Sterile,1/pk, 12/cs. Ideal for shaker culture applications. There are two kinds of bottle materials, both PETG and PC are available. PC bottle or PEFG bottleSterilized by E-BeamNonpyrogenicIndividually wrapped in a plastic bag.Both vent caps and plug seal caps are available.Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.